missing translation for 'onlineSavingsMsg'
Learn More

1-(3-Bromo-2-fluorophenyl)ethanol, 98%, Thermo Scientific™

Catalog Number 15422049
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
1221715-80-5
C8H8BrFO
219.05
MFCD16140247
QFVPIDFXSBXICP-UHFFFAOYNA-N
1-3-bromo-2-fluorophenyl ethanol, 1-3-bromo-2-fluoro-phenyl ethanol, 1-3-bromo-2-fluorophenyl ethan-1-ol, 3-bromo-2-fluoro-alpha-methylbenzenemethanol
53484284
1-(3-bromo-2-fluorophenyl)ethanol
CC(O)C1=C(F)C(Br)=CC=C1
This item is not returnable. View return policy
1221715-80-5
C8H8BrFO
219.05
MFCD16140247
QFVPIDFXSBXICP-UHFFFAOYNA-N
1-3-bromo-2-fluorophenyl ethanol, 1-3-bromo-2-fluoro-phenyl ethanol, 1-3-bromo-2-fluorophenyl ethan-1-ol, 3-bromo-2-fluoro-alpha-methylbenzenemethanol
53484284
1-(3-bromo-2-fluorophenyl)ethanol
CC(O)C1=C(F)C(Br)=CC=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 219.05
Percent Purity 98%
Chemical Name or Material 1-(3-Bromo-2-fluorophenyl)ethanol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'