Learn More
1,3-Bis(aminopropyl)tetramethyldisiloxane, 94%
Description
1,3-Bis(aminopropyl)tetramethyldisiloxane is used as a curing agent for epoxy molding compounds, which is utilized for high-reliability semiconductor devices. It is also used to prepare siloxane- urea copymers from 4,4’-diphenylmethane diisocyanate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
1,3-Bis(aminopropyl)tetramethyldisiloxane is used as a curing agent for epoxy molding compounds, which is utilized for high-reliability semiconductor devices. It is also used to prepare siloxane- urea copymers from 4,4’-diphenylmethane diisocyanate.
Solubility
Slightly soluble in water.
Notes
Air Sensitive. Incompatible with strong oxidizing agents. Store in a cool place.
Specifications
Specifications
| Boiling Point | 132°C to 139°C (11mmHg) |
| Quantity | 100 g |
| UN Number | UN2735 |
| Beilstein | 1760876 |
| Flash Point | 91°C (195°F) |
| Solubility Information | Slightly soluble in water. |
| Formula Weight | 248.52 |
| Percent Purity | 94% |
| Refractive Index | 1.45 |
| Sensitivity | Air Sensitive |
| Show More |
Safety and Handling
- 1,3-Bis(aminopropyl)tetramethyldisiloxane
- Danger
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 C
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H412-Harmful to aquatic life with long lasting effects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : 1,3-Bis(aminopropyl)tetramethyldisiloxane
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.