Learn More
1,3,7-Trichloronaphthalene, TRC
Specifications
Specifications
| Chemical Name or Material | 1,3,7-Trichloronaphthalene |
| CAS | 55720-37-1 |
| Molecular Formula | C10 H5 Cl3 |
| Quantity | 25 mg |
| Synonym | PCN 21 (1,3,7-Trichloronaphthalene) |
| InChI Formula | InChI=1S/C10H5Cl3/c11-7-2-1-6-3-8(12)5-10(13)9(6)4-7/h1-5H |
| SMILES | Clc1ccc2cc(Cl)cc(Cl)c2c1 |
| IUPAC Name | 1,3,7-trichloronaphthalene |
| Molecular Weight (g/mol) | 231.51 |
| Formula Weight | 229.9457 |