missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Trimethoxy-2-nitrobenzene, 98%

Product Code. 11478703
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11478703

Brand: Thermo Scientific Alfa Aesar A10473.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14227-18-0 | C9H11NO5 | 213.189 g/mol

2,4,6-Trimethoxynitrobenzene is used as a chemicals, pharmaceutical intermediate, chemical research, application intermediate, pharmaceutical reagents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4,6-Trimethoxynitrobenzene is used as a chemicals, pharmaceutical intermediate, chemical research, application intermediate, pharmaceutical reagents.

Solubility
Soluble in water.

Notes
Desiccate at room temperature. Incompatabile to strong oxidizing agents, strong acids and bases. Store in a cool, dry, well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14227-18-0
Molecular Formula C9H11NO5
Molecular Weight (g/mol) 213.189
MDL Number MFCD00016992
InChI Key VWYAWLZEMLQGJH-UHFFFAOYSA-N
PubChem CID 518903
IUPAC Name 1,3,5-trimethoxy-2-nitrobenzene
SMILES COC1=CC(=C(C(=C1)OC)[N+](=O)[O-])OC

Specifications

Melting Point 150°C to 153°C
Quantity 25 g
Beilstein 2277262
Synonym 2,4,6-trimethoxynitrobenzene, 1-nitro-2,4,6-trimethoxybenzene, benzene,1,3,5-trimethoxy-2-nitro, benzene, 1,3,5-trimethoxy-2-nitro, acmc-20ap5l, maybridge1_001624, 1,3,5-trimethoxy-2-nitro-benzene, #
Solubility Information Soluble in water.
Formula Weight 213.19
Percent Purity 98%
Chemical Name or Material 1,3,5-Trimethoxy-2-nitrobenzene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.