Learn More
1,3,5-O-Methylidyne-myo-inositol, TRC
Product Code. 30477060
Change view
Click to view available options
Quantity:
2.5 g
25 g
Unit Size:
2.5g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30477060 25 g 25g
30473237 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30477060 Supplier Toronto Research Chemicals Supplier No. TRCM31294025G
 Restricted Item
Estimated Shipment: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 98510-20-4
Molecular Formula C7H10O6
Molecular Weight (g/mol) 190.15
Synonym 2,4,10-Trioxatricyclo[3.3.1.13,7]decane, myo-Inositol deriv., myo-Inositol-1,3,5-orthoformate, myo-Inositol Monoorthoformate
SMILES O[C@@H]1[C@H]2O[C@@H]3OC([C@@H](O)C1O3)[C@@H]2O
Specifications
Quantity 25 g
InChI Formula InChI=1S/C7H10O6/c8-1-4-2(9)6-3(10)5(1)12-7(11-4)13-6/h1-10H/t1-,2-,3-,4+,5?,6?,7-/m0/s1
Formula Weight 190.0477
Chemical Name or Material 1,3,5-O-Methylidyne-myo-inositol
Produkttitel
Vælg et problem
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.