missing translation for 'onlineSavingsMsg'
Learn More

1-[3,5-Bis(trifluoromethyl)phenyl]pyrrole, 97%, Thermo Scientific™

Catalog Number 11472408
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
175136-60-4
C12H7F6N
279.185
MFCD00053048
BKSCRBOUGNMEKQ-UHFFFAOYSA-N
1-3,5-bis trifluoromethyl phenyl pyrrole, 1-3,5-bis trifluoromethyl phenyl-1h-pyrrole, pubchem9334, maybridge1_002820, acmc-1c4qv, 3,5-bis trifluoromethyl phenyl pyrrole, 1-3,5-di trifluoromethyl phenyl-1h-pyrrole
2736181
1-[3,5-bis(trifluoromethyl)phenyl]pyrrole
C1=CN(C=C1)C2=CC(=CC(=C2)C(F)(F)F)C(F)(F)F
This item is not returnable. View return policy
175136-60-4
C12H7F6N
279.185
MFCD00053048
BKSCRBOUGNMEKQ-UHFFFAOYSA-N
1-3,5-bis trifluoromethyl phenyl pyrrole, 1-3,5-bis trifluoromethyl phenyl-1h-pyrrole, pubchem9334, maybridge1_002820, acmc-1c4qv, 3,5-bis trifluoromethyl phenyl pyrrole, 1-3,5-di trifluoromethyl phenyl-1h-pyrrole
2736181
1-[3,5-bis(trifluoromethyl)phenyl]pyrrole
C1=CN(C=C1)C2=CC(=CC(=C2)C(F)(F)F)C(F)(F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 41°C to 43°C
Quantity 1 g
Formula Weight 279.19
Percent Purity 97%
Chemical Name or Material 1-[3,5-Bis(trifluoromethyl)phenyl]pyrrole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'