Learn More
1,3,5-Benzene-2,4,6-d3-tricarboxylic Acid, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | Trimesic Acid-D3 |
| CAS | 62790-27-6 |
| Molecular Formula | C9 D3 H3 O6 |
| Quantity | 0.5 g |
| Synonym | 1,3,5-Benzene-2,4,6-d3-tricarboxylic acid (9 CI), 1,3,5-Benzene-2,4,6-d3-tricarboxylic Acid, 1,3,5-Benzenetricarboxylic Acid-2,4,6-d3, 1,3,5-Benzenetricarboxylic Acid-2,4,6-D3, Trimesic Acid-D3, 1,3,5-Tricarboxybenzene-2,4,6-d3, 5-Carboxyisophthalic acid-d3, Trimesinic acid-d3, Trimesitinic acid-d3 |
| InChI Formula | InChI=1 S/C9H6O6/c10-7(11)4-1-5(8(12)13)3-6(2-4)9(14)15/h1-3 H,(H,10,11)(H,12,13)(H,14,15)/i1D,2 D,3 D |
| SMILES | [2 H]c1c(C(=O)O)c([2 H])c(C(=O)O)c([2 H])c1C(=O)O |
| IUPAC Name | 2,4,6-trideuteriobenzene-1,3,5-tricarboxylic acid |
| Molecular Weight (g/mol) | 213.1588 |
| Formula Weight | 213.0353 g/mol |
| Show More |