Learn More
1-(3,4-Dihydroxyphenyl)-2-(methylamino)ethanone Hydrochloride (Adrenalone Hydrochloride), Mikromol™
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
1-(3,4-Dihydroxyphenyl)-2-(methylamino)ethanone Hydrochloride (Adrenalone Hydrochloride)
Specifications
Specifications
| Chemical Name or Material | 1-(3,4-Dihydroxyphenyl)-2-(methylamino)ethanone Hydrochloride (Adrenalone Hydrochloride) |
| CAS | 62-13-5 |
| Analyte or Component Names | 1-(3,4-Dihydroxyphenyl)-2-(methylamino)ethanone Hydrochloride (Adrenalone Hydrochloride) |
| Shipping Condition | Room Temperature |
| Molecular Formula | C9 H11 N O3 . Cl H |
| Certifications/Compliance | ISO 17034 |
| Synonym | Adrenaline Tartrate Imp. C (EP), Adrenaline Imp. C (EP), Adrenalone Hydrochloride, 1-(3,4-Dihydroxyphenyl)-2-(methylamino)ethanone Hydrochloride, Adrenaline Tartrate Imp. C (EP) as Hydrochloride, Ethanone, 1-(3,4-dihydroxyphenyl)-2-(methylamino)-, hydrochloride (1:1), Acetophenone, 3',4'-dihydroxy-2-(methylamino)-, hydrochloride (7CI,8CI), Adrenalone hydrochloride (6CI), Ethanone, 1-(3,4-dihydroxyphenyl)-2-(methylamino)-, hydrochloride (9CI), Adrenone hydrochloride, Kephrine hydrochloride, Styphnonasal, alpha-Methylamino-3,4-dihydroxyacetophenone hydrochloride, alpha-Methylamino-3',4'-dihydroxyacetophenone hydrochloride, Dipivefrin Related Compound B as Hydrochloride, Adrenaline Tartrate Impurity C as Hydrochloride, Dipivefrine Hydrochloride Impurity B as Hydrochloride, Adrenaline Impurity C as Hydrochloride, Dipivefrine Impurity B as Hydrochloride |
| InChI Formula | InChI=1S/C9H11NO3.ClH/c1-10-5-9(13)6-2-3-7(11)8(12)4-6;/h2-4,10-12H,5H2,1H3;1H |
| SMILES | Cl.CNCC(=O)c1ccc(O)c(O)c1 |
| IUPAC Name | 1-(3,4-dihydroxyphenyl)-2-(methylamino)ethanone;hydrochloride |
| Show More |