Learn More
1,3,2-Dioxathiolane 2,2-Dioxide, TRC
Product Code. 30369907
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30369907 5 g
30377351 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30369907 Supplier Toronto Research Chemicals Supplier No. TRCD4868655G
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1072-53-3
Molecular Formula C2 H4 O4 S
Molecular Weight (g/mol) 124.12
Synonym Ethylene Glycol Cyclic Sulfate, 1,2-Ethylene Sulfate, Ethylene Sulfate, NSC 526594
IUPAC Name 1,3,2-dioxathiolane 2,2-dioxide
SMILES O=S1(=O)OCCO1
Specifications
Quantity 5 g
InChI Formula InChI=1S/C2H4O4S/c3-7(4)5-1-2-6-7/h1-2H2
Formula Weight 123.983
Chemical Name or Material 1,3,2-Dioxathiolane 2,2-Dioxide