Learn More
1-[(2RS)-2-(2,4-Dichlorophenyl)-2-[(2,5-dichlorothiophen-3-yl)methoxy]ethyl]-1H-imidazole Hydrochloride, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
1-[(2RS)-2-(2,4-Dichlorophenyl)-2-[(2,5-dichlorothiophen-3-yl)methoxy]ethyl]-1H-imidazole Hydrochloride
Specifications
Specifications
| Chemical Name or Material | 1-[(2RS)-2-(2,4-Dichlorophenyl)-2-[(2,5-dichlorothiophen-3-yl)methoxy]ethyl]-1H-imidazole Hydrochloride |
| CAS | 61675-62-5 |
| Analyte or Component Names | 1-[(2RS)-2-(2,4-Dichlorophenyl)-2-[(2,5-dichlorothiophen-3-yl)methoxy]ethyl]-1H-imidazole Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C16 H12 Cl4 N2 O S . Cl H |
| Certifications/Compliance | ISO 17025 |
| Synonym | 1-[2-(2,4-Dichlorophenyl)-2-[(2,5-dichloro-3-thienyl)methoxy]ethyl]-1H-imidazole Hydrochloride (1:1), 1H-Imidazole, 1-[2-(2,4-dichlorophenyl)-2-[(2,5-dichloro-3-thienyl)methoxy]ethyl]-, monohydrochloride (9CI), Tioconazole Related Compound B, Tioconazole Related Compound B as Hydrochloride, Tioconazole Impurity B as Hydrochloride |
| InChI Formula | InChI=1S/C16H12Cl4N2OS.ClH/c17-11-1-2-12(13(18)6-11)14(7-22-4-3-21-9-22)23-8-10-5-15(19)24-16(10)20;/h1-6,9,14H,7-8H2;1H |
| SMILES | Cl.Clc1ccc(C(Cn2ccnc2)OCc3cc(Cl)sc3Cl)c(Cl)c1 |
| IUPAC Name | 1-[2-(2,4-dichlorophenyl)-2-[(2,5-dichlorothiophen-3-yl)methoxy]ethyl]imidazole;hydrochloride |
| Show More |