missing translation for 'onlineSavingsMsg'
Learn More

1,2-Propanediol, Supelco™

Product Code. 15881791
Change view
Click to view available options
Quantity:
1 L
5 L
Unit Size:
1 L
5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15881791 5 L 5 L
15871791 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15881791 Supplier Supelco Supplier No. 8223245000
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

EMPLURA™

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57-55-6
Grade Research
Molecular Formula CH3CH(OH)CH2OH
Molecular Weight (g/mol) 76.09

Specifications

Melting Point -59°C
Density 1.04 g/cm3 at 20°C
Boiling Point 188°C/1013 hPa
MDL Number MFCD00064272
Quantity 5 L
Synonym 1,2-Propanediol, 1,2-Propylene glycol, 1,2-Propanediol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.