Learn More
1-(2-(Phenanthren-9-yl)phenyl)propan-2-one, TRC
Specifications
Specifications
| Chemical Name or Material | 1-(2-(Phenanthren-9-yl)phenyl)propan-2-one |
| CAS | 1013933-56-6 |
| Molecular Formula | C23H18O |
| Quantity | 10 mg |
| InChI Formula | InChI=1S/C23H18O/c1-16(24)14-17-8-2-5-11-20(17)23-15-18-9-3-4-10-19(18)21-12-6-7-13-22(21)23/h2-13,15H,14H2,1H3 |
| SMILES | CC(=O)Cc1ccccc1c2cc3ccccc3c4ccccc24 |
| IUPAC Name | 1-(2-phenanthren-9-ylphenyl)propan-2-one |
| Molecular Weight (g/mol) | 310.39 |
| Formula Weight | 310.1358 |