Learn More
1-(2-Hydroxyphenyl)-3-phenyl-2-propenone, Sigma-Aldrich™
Specifications
Specifications
| CAS | 1214-47-7 |
| Melting Point | 87°C |
| MDL Number | MFCD00016441 |
| Quantity | 25 g |
| Synonym | 1-(2-Hydroxyphenyl)-3-phenyl-2-propenone, 1-(2-Hydroxyphenyl)-3-phenyl-2-propenone |
| Molecular Weight (g/mol) | 224.25 |
| Grade | Research |