Learn More
1,2-Epoxy-7-octene, 97%
Product Code. p-7058318
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11327106 5 g 5g
11337106 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11327106 Supplier Thermo Scientific Alfa Aesar Supplier No. L07033.06
Request Bulk or Custom Format
CAS: 19600-63-6 | C8H14O | 126.199 g/mol
1,2-Epoxy-7-octene is used as an organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,2-Epoxy-7-octene is used as an organic chemical synthesis intermediate.
Solubility
Not miscible in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 19600-63-6
Molecular Formula C8H14O
Molecular Weight (g/mol) 126.199
MDL Number MFCD00005156
InChI Key UKTHULMXFLCNAV-UHFFFAOYSA-N
Synonym 1,2-epoxy-7-octene, oxirane, 5-hexenyl, 5-hexenyloxirane, 7,8-epoxyoctene, ccris 3749, 2-hex-5-en-1-yl oxirane, oxirane, 5-hexenyl-, s, 2-hex-5-en-yloxirane, 2-5-hexenyl oxirane, acmc-20ap64
PubChem CID 29678
IUPAC Name 2-hex-5-enyloxirane
SMILES C=CCCCCC1CO1
Specifications
Density 0.85
Boiling Point 167°C
Flash Point 38°C (100°F)
Refractive Index 1.435
Quantity 5 g
UN Number UN1993
Beilstein 1305290
Solubility Information Not miscible in water.
Formula Weight 126.2
Percent Purity 97%
Chemical Name or Material 1,2-Epoxy-7-octene