Learn More
1,2-Dichlorobenzene, Sigma-Aldrich™
Product Code. 15858960
Change view
Click to view available options
Quantity:
100 mL
1 L
2.5 L
Unit Size:
1 L
100mL
2.5 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15858960 2.5 L 2.5 L
15826360 100 mL 100mL
15848960 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15858960 Supplier Sigma Aldrich Supplier No. 8032382500
 Restricted Item
Estimated Shipment: 18-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 95-50-1
Grade Research
Molecular Formula 1,2-(Cl)2C6H4
Molecular Weight (g/mol) 147
Specifications
Melting Point -17°C
Density 1.31 g/cm3 at 20°C
Boiling Point 180°C/1013 hPa
MDL Number MFCD00000535
Quantity 2.5 L
Synonym 1,2-Dichlorobenzene, o-Chlorobenzene, 1,2-Dichlorobenzene