Learn More
1-(2’-Deoxy-3’,5’-di-O-toluoyl-Beta-D-ribofuranosyl)-2-oxo-4-amino-1,2-dihydro-1,3,5-triazine, TRC
Specifications
Specifications
| Chemical Name or Material | 1-(2'-Deoxy-3',5'-di-O-toluoyl-beta-D-ribofuranosyl)-2-oxo-4-amino-1,2-dihydro-1,3,5-triazine |
| CAS | 10302-79-1 |
| Molecular Formula | C24 H24 N4 O6 |
| Quantity | 5 mg |
| Synonym | 1,3,5-Triazin-2(1H)-one, 4-amino-1-[2-deoxy-3,5-bis-O-(4-methylbenzoyl)-β-D-erythro-pentofuranosyl]- (9CI, ACI), 1,3,5-Triazin-2(1H)-one, 4-amino-1-(2-deoxy-β-D-erythro-pentofuranosyl)-, bis(4-methylbenzoate) (ester) (ZCI), 4-Amino-1-[2-deoxy-3,5-bis-O-(4-methylbenzoyl)-β-D-erythro-pentofuranosyl]-1,3,5-triazin-2(1H)-one (ACI), s-Triazin-2(1H)-one, 4-amino-1-(2-deoxy-β-D-erythro-pentofuranosyl)-, di-p-toluate (7CI), s-Triazin-2(1H)-one, 4-amino-1-(2-deoxy-β-D-erythro-pentofuranosyl)-, di-p-toluate (ester) (8CI), 1-(2'-Desoxy-3',5'-di-O-p-toluoyl-β-D-ribofuranosyl)-2-oxo-4-amino-1,2-dihydro-1,3,5-triazine, 3',5'-Di-O-(4-methylbenzoyl)decitabine |
| InChI Formula | InChI=1S/C24H24N4O6/c1-14-7-3-5-9-16(14)21(29)32-12-19-18(34-22(30)17-10-6-4-8-15(17)2)11-20(33-19)28-13-26-23(25)27-24(28)31/h3-10,13,18-20H,11-12H2,1-2H3,(H2,25,27,31)/t18-,19+,20+/m0/s1 |
| SMILES | Cc1ccccc1C(=O)OC[C@H]2O[C@H](C[C@@H]2OC(=O)c3ccccc3C)N4C=NC(=NC4=O)N |
| IUPAC Name | [(2R,3S,5R)-5-(4-amino-2-oxo-1,3,5-triazin-1-yl)-3-(2-methylbenzoyl)oxyoxolan-2-yl]methyl 2-methylbenzoate |
| Molecular Weight (g/mol) | 464.47 |
| Formula Weight | 464.1696 |