missing translation for 'onlineSavingsMsg'
Learn More

1,2-Bis(dimethylphosphino)ethane, 97%, Thermo Scientific Chemicals

Product Code. p-7203420 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
500 mg
1 g
5 g
Unit Size:
1g
500mg
5g
This item is not returnable. View return policy

Product Code. 10482451

Brand: Thermo Scientific Acros 310035000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23936-60-9
Molecular Formula C6H16P2
Molecular Weight (g/mol) 150.14
MDL Number MFCD00008511
InChI Key ZKWQSBFSGZJNFP-UHFFFAOYSA-N
Synonym acmc-1cjp0, zkwqsbfsgzjnfp-uhfffaoysa, dimethyl-2-dimethylphosphino ethylphosphine, 1,2-bis dimethylphosphino ethane, 2-dimethylphosphanyl ethyl dimethylphosphane, 2-dimethylphosphanylethyl dimethyl phosphane, ethylenebis dimethylphosphine, pubchem6482, ghl.pd_mitscher_leg0.173, phosphine, 1,2-ethanediylbis dimethyl
PubChem CID 141059
SMILES CP(C)CCP(C)C

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless
Density 0.9 g/mL
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula (CH3)PCHCHP(CH3)
Refractive Index 1.5060 to 1.5080 (20°C, 589 nm)
Quantity 500 mg
IUPAC Name [2-(dimethylphosphanyl)ethyl]dimethylphosphane
Formula Weight 150.14
Percent Purity ≥96.0%
Physical Form Liquid
Chemical Name or Material 1, 2-Bis(dimethylphosphino)ethane
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.