Learn More
1,2,4-Trichlorobenzene, Sigma-Aldrich™
Product Code. 15814611
Change view
Click to view available options
Quantity:
1 L
2.5 L
Unit Size:
1 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15814611 2.5 L 2.5 L
15804611 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 15814611 Supplier Sigma Aldrich Supplier No. 8211522500
 Restricted Item
Estimated Shipment: 18-11-2026
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 120-82-1
Grade Research
Molecular Formula C6H3-1,2,4-(Cl)3
Molecular Weight (g/mol) 181.45
Specifications
Melting Point 17°C
Density 1.45 g/cm3 at 20°C
Quantity 2.5 L
MDL Number MFCD00000547
Synonym 1,2,4-Trichlorobenzene, 1,2,4-Trichlorobenzene
Boiling Point 213.5°C/1013 hPa
Chemical Name or Material 1,2,4-Trichlorobenzene