Learn More
1,2,4-Triazole, Sigma-Aldrich™
Specifications
Specifications
| CAS | 288-88-0 |
| Melting Point | 119°C to 122°C |
| Density | 1.15 g/cm3 at 130°C |
| Boiling Point | 259°C to 261°C/1013 hPa |
| MDL Number | MFCD00005228 |
| Quantity | 25 g |
| Synonym | 1,2,4-Triazole, 1,2,4-Triazole |
| Molecular Weight (g/mol) | 69.07 |
| Grade | Research |