Learn More
1,2,4-Benzenetricarboxylic 1,2-Anhydride, Sigma-Aldrich™
Product Code. 15860771
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15860771 500 g 500g
15850771 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15860771 Supplier Sigma-Aldrich Supplier No. 8201380500
 Restricted Item
Estimated Shipment: 09-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 552-30-7
Grade Research
Molecular Weight (g/mol) 192.13
Specifications
Melting Point 164°C to 168°C
Density 1.487 g/cm3 at 20°C
Boiling Point 240°C/19 hPa
MDL Number MFCD00005925
Quantity 500 g
Synonym 1,2,4-Benzenetricarboxylic 1,2-anhydride, Trimellitic acid anhydride, 1,2,4-Benzenetricarboxylic 1,2-anhydride