Learn More
1,2,3-O-Tris(triisopropyl) Glycerol-d5, TRC
Specifications
Specifications
| Chemical Name or Material | 1,2,3-O-Tris(triisopropyl) Glycerol-d5 |
| CAS | 1330185-53-9 |
| Molecular Formula | C30H63D5O3Si3 |
| Quantity | 10 mg |
| Synonym | 1,2,3-O-Tris(triisopropyl) 1,2,3-Propanetriol-d5, 1,2,3-O-Tris(triisopropyl) 1,3-Dihydroxy-2-propanol-d5, 1,2,3-O-Tris(triisopropyl) Propanetriol-d5, 1,2,3-O-Tris(triisopropyl) 1,2,3-Propane-1,1,2,3,3-d5-triol, 1,2,3-O-Tris(triisopropyl) 1,2,3-Trihydroxypropane-d5, 1,2,3-O-Tris(triisopropyl) Bulbold-d5, 1,2,3-O-Tris(triisopropyl) Cognis G-d5, 1,2,3-O-Tris(triisopropyl) Cristal-d5, 1,2,3-O-Tris(triisopropyl) DG Glycerin-d5, 1,2,3-O-Tris(triisopropyl) Trihydroxypropane-d5 |
| InChI Formula | InChI=1S/C30H68O3Si3/c1-21(2)34(22(3)4,23(5)6)31-19-30(33-36(27(13)14,28(15)16)29(17)18)20-32-35(24(7)8,25(9)10)26(11)12/h21-30H,19-20H2,1-18H3/i19D2,20D2,30D |
| SMILES | [2H]C([2H])(O[Si](C(C)C)(C(C)C)C(C)C)C([2H])(O[Si](C(C)C)(C(C)C)C(C)C)C([2H])([2H])O[Si](C(C)C)(C(C)C)C(C)C |
| IUPAC Name | [1,1,2,3,3-pentadeuterio-1,3-bis[tri(propan-2-yl)silyloxy]propan-2-yl]oxy-tri(propan-2-yl)silane |
| Molecular Weight (g/mol) | 566.15 |
| Formula Weight | 565.479 |