Learn More
1,2,3,6-Tetrahydro-Alpha,4-bis(p-methoxyphenyl)-1-methyl-3-pyridinemethanol, TRC
Specifications
Specifications
| Chemical Name or Material | 1,2,3,6-Tetrahydro-alpha,4-bis(p-methoxyphenyl)-1-methyl-3-pyridinemethanol |
| CAS | 10185-04-3 |
| Molecular Formula | C21H25NO3 |
| Quantity | 25 mg |
| Synonym | 1,2,3,6-Tetrahydro-α,4-bis(4-methoxyphenyl)-1-methyl-3-pyridinemethanol |
| InChI Formula | InChI=1S/C21H25NO3/c1-22-13-12-19(15-4-8-17(24-2)9-5-15)20(14-22)21(23)16-6-10-18(25-3)11-7-16/h4-12,20-21,23H,13-14H2,1-3H3 |
| SMILES | OC(C1C(C2=CC=C(OC)C=C2)=CCN(C1)C)C3=CC=C(C=C3)OC |
| IUPAC Name | (4-methoxyphenyl)(4-(4-methoxyphenyl)-1-methyl-1,2,3,6-tetrahydropyridin-3-yl)methanol |
| Molecular Weight (g/mol) | 399.44 |
| Formula Weight | 399.18 |