Learn More
1,2,3,4-Tetrahydro-1-napthol, TRC
Specifications
Specifications
| Chemical Name or Material | 1,2,3,4-Tetrahydro-1-napthol |
| CAS | 529-33-9 |
| Molecular Formula | C10H20O |
| Quantity | 5 g |
| Synonym | (RS)-1-Hydroxytetralin, (±)-1,2,3,4-Tetrahydro-1-naphthol, (±)-1-Tetralol, (±)-alpha-Tetralol1,2,3,4-Tetrahydro-1-naphthalenol, 1,2,3,4-Tetrahydro-1-naphthol, 1-Hydroxy-1,2,3,4-tetrahydronaphthalene, 1-Hydroxytetralin, 1-Tetralol, NSC 5172alpha-Tetralol |
| InChI Formula | InChI=1S/C10H12O/c11-10-7-3-5-8-4-1-2-6-9(8)10/h1-2,4,6,10-11H,3,5,7H2 |
| SMILES | OC1CCCc2ccccc12 |
| IUPAC Name | 1,2,3,4-tetrahydronaphthalen-1-ol |
| Molecular Weight (g/mol) | 148.2 |
| Formula Weight | 148.0888 |