Learn More
1,2,3,4-tetrahydro-1,2-dimethyl-4,8-isoquinolinediol, TRC
Product Code. 30419809
Change view
Click to view available options
Quantity:
2.5 mg
Unit Size:
2.5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30419809 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30419809 Supplier Toronto Research Chemicals Supplier No. TRCT2934902.5MG
 Restricted Item
Estimated Shipment: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1,2,3,4-tetrahydro-1,2-dimethyl-4,8-isoquinolinediol
CAS 102830-20-6
Molecular Formula C11 H15 N O2
Quantity 2.5 mg
InChI Formula InChI=1S/C11H15NO2/c1-7-11-8(4-3-5-9(11)13)10(14)6-12(7)2/h3-5,7,10,13-14H,6H2,1-2H3
SMILES CC1N(C)CC(O)c2cccc(O)c12
IUPAC Name 1,2-dimethyl-3,4-dihydro-1H-isoquinoline-4,8-diol
Molecular Weight (g/mol) 193.242
Formula Weight 193.11