missing translation for 'onlineSavingsMsg'
Learn More

1,2,3,4-Butanetetracarboxylic acid, 98+%

Product Code. p-7023252
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
Product Code. Quantity unitSize
11447957 100 g 100g
11457957 500 g 500g
2 options
This item is not returnable. View return policy

Product Code. 11447957

Brand: Thermo Scientific Alfa Aesar A16612.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1703-58-8 | C8H10O8 | 234.16 g/mol

1,2,3,4-Butanetetracarboxylic acid is used as a cross linking agent to prepare cotton cellulose in order to improve anti-prilling and flame retardant properties. It is used as a spacer in the cross-linking of titania nanoparticles to cotton. Further, it is used as a nonformaldehyde durable press finishing agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2,3,4-Butanetetracarboxylic acid is used as a cross linking agent to prepare cotton cellulose in order to improve anti-prilling and flame retardant properties. It is used as a spacer in the cross-linking of titania nanoparticles to cotton. Further, it is used as a nonformaldehyde durable press finishing agent.

Solubility
Soluble in water and ethanol.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1703-58-8
Molecular Formula C8H10O8
Molecular Weight (g/mol) 234.16
MDL Number MFCD00002722
InChI Key GGAUUQHSCNMCAU-UHFFFAOYSA-N
Synonym 1,2,3,4-butanetetracarboxylic acid, butanetetracarboxylic acid, dsstox_cid_4670, dsstox_rid_77494, dsstox_gsid_24670, 1,2,3,4-butanetetracarboxylic, meso-1,2,3,4-butanetetracarboxylic acid, acmc-20abw3, ksc176c1b, ggauuqhscnmcau-uhfffaoysa
PubChem CID 15560
IUPAC Name butane-1,2,3,4-tetracarboxylic acid
SMILES C(C(C(CC(=O)O)C(=O)O)C(=O)O)C(=O)O

Specifications

Melting Point ∼190°C to 195°C (decomposition)
Quantity 100 g
Beilstein 1729167
Solubility Information Soluble in water and ethanol.
Formula Weight 234.16
Percent Purity ≥98%
Chemical Name or Material 1,2,3,4-Butanetetracarboxylic acid
compliance-icons compliance-icons
Product Identifier
  • 1,2,3,4-Butanetetracarboxylic acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : 1,2,3,4-Butanetetracarboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.