missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 1,2,3,4,6-Penta-O-acetyl-D-mannopyranose, 98%

Product Code. 15448559
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 15448559

Brand: Thermo Scientific Chemicals J66973.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 25941-03-1 | C16H22O11 | 390.341 g/mol

1,2,3,4,6-Penta-O-acetyl-D-mannopyranose is used for the preparation of glycopyranoside phosphates for the study of T-lymphocyte mediated inflammatory diseases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2,3,4,6-Penta-O-acetyl-D-mannopyranose is used for the preparation of glycopyranoside phosphates for the study of T-lymphocyte mediated inflammatory diseases.

Solubility
Soluble in dichloromethane, ether, ethyl acetate, methanol, and chloroform.

Notes
Keep container tightly closed in a dry and well-ventilated place. Store in cool place. Store at -20°C.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25941-03-1
Molecular Formula C16H22O11
Molecular Weight (g/mol) 390.341
MDL Number MFCD05864874
InChI Key LPTITAGPBXDDGR-WHWZVRATSA-N
Synonym 1,2,3,4,6-penta-o-acetyl-d-mannopyranose, d-mannopyranose pentaacetate, 3s,4s,5r,6r-6-acetoxymethyl tetrahydro-2h-pyran-2,3,4,5-tetrayl tetraacetate, 2r,3r,4s,5s-3,4,5,6-tetrakis acetyloxy oxan-2-yl methyl acetate, mannopyranose pentaacetate, d-mannopyranose, pentaacetate, 1,2,3,4,6-pentaacetate d-mannopyranose, d-mannopyranose,1,2,3,4,6-pentaacetate, 1,2,3,4,6-penta-o-acetyl-dmannopyranose, 1,2,3,4,6-penta-o-acetyl-alpha,beta-d-mannose
PubChem CID 11811209
IUPAC Name [(2R,3R,4S,5S)-3,4,5,6-tetraacetyloxyoxan-2-yl]methyl acetate
SMILES CC(=O)OCC1C(C(C(C(O1)OC(=O)C)OC(=O)C)OC(=O)C)OC(=O)C

Specifications

Melting Point 66°C to 72°C
Color White
Quantity 25 g
Solubility Information Soluble in dichloromethane,ether,ethyl acetate,methanol,and chloroform.
Optical Rotation +54° (c=1 in Chloroform)
Formula Weight 390.34
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 1,2,3,4,6-Penta-O-acetyl-D-mannopyranose

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.