Learn More
Trajan Scientific and Medical™ 1/16 in. Graphite/Vespel™ Fittings Reducing Ferrule
Made from 15% Graphite and 85% Vespel
Brand: Trajan Scientific and Medical™ 072658
Additional Details : Weight : 1.00000kg
Description
- Composite of graphite and Vespel
- Mechanically robust, hard material, long lifetime
- Forms strong grip with capillary column
- Hole I.D. smaller than nominal size of fitting
Specifications
3.2 mm | |
3.7 mm | |
0.9 mm |
15% Graphite, 85% Vespel | |
Graphite Vespel Reducing Ferrules 1/16 in. Fitting 0.9mm x 3.2mm x 3.7mm |