Learn More
1,10-Phenanthroline, Sigma-Aldrich™
Specifications
Specifications
| CAS | 66-71-7 |
| Melting Point | 117°C to 119°C |
| MDL Number | MFCD00011678 |
| Quantity | 5 g |
| Synonym | 1,10-Phenanthroline, 1,10-Phenanthroline |
| Molecular Weight (g/mol) | 180.21 |
| Grade | Research |