Learn More
1,1,1-Trichloroacetone, SPEX CertiPrep™
Product Code. 16320208 Shop All SPEX Certiprep Products
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Product Code. 16320208
Brand: SPEX Certiprep S3577
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Includes
Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
AnalyteCAS No.Concentration
1,1,1-Trichloroacetone918-00-31000μg/mL
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Packaging 2mL amber prescored glass ampule
CAS 918-00-3, 67-56-1
CAS Min % 0.1, 99.9
Concentration or Composition Notes Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
CAS Max % 0.1, 99.9
Molecular Formula C3H3Cl3O
MDL Number MFCD00018829
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
InChI Key SMZHKGXSEAGRTI-UHFFFAOYSA-N
SMILES CC(=O)C(Cl)(Cl)Cl
IUPAC Name 1,1,1-trichloropropan-2-one
Solvent or Matrix Methanol-P&T
Molecular Weight (g/mol) 161.41
PubChem CID 13514
Quantity 1 mL
Chemical Name or Material 1,1,1-Trichloroacetone
Show More Show Less
Product intended for laboratory use only