Learn More
1,1,1,3,3,3-Hexafluoroisopropyl methacrylate, 99%, stab.
Product Code. 11489437
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
Product Code. Quantity unitSize
11489437 25 g 25g
11469437 1 g 1g
11479437 5 g 5g
3 options
Product Code. 11489437
Brand: Thermo Scientific Alfa Aesar B21447.14
Request Bulk or Custom Format
CAS: 3063-94-3 | C7H6F6O2 | 236.113 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 3063-94-3
Molecular Formula C7H6F6O2
Molecular Weight (g/mol) 236.113
MDL Number MFCD00040105
InChI Key FMQPBWHSNCRVQJ-UHFFFAOYSA-N
Synonym hexafluoroisopropyl methacrylate, 1,1,1,3,3,3-hexafluoroisopropyl methacrylate, 2-propenoic acid, 2-methyl-, 2,2,2-trifluoro-1-trifluoromethyl ethyl ester, methacrylic acid 1,1,1,3,3,3-hexafluoroisopropyl ester, 2,2,2-trifluoro-1-trifluoromethyl ethyl methacrylate, hexafluoro-2-propyl methacrylate, 1h-1-trifluoromethyl trifluoroethyl methacrylate, 1,1,1,3,3,3-hexafluoroprop-2-yl 2-methylprop-2-enoate, 1,1,1,3,3,3-hexafluoro-2-propanyl methacrylate, hfipma
PubChem CID 76469
IUPAC Name 1,1,1,3,3,3-hexafluoropropan-2-yl 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OC(C(F)(F)F)C(F)(F)F
Specifications
Density 1.302
Boiling Point 99°C
Flash Point 14°C (57°F)
Refractive Index 1.331
Quantity 25 g
UN Number UN1993
Formula Weight 236.12
Percent Purity 99%
Chemical Name or Material 1,1,1,3,3,3-Hexafluoroisopropyl methacrylate, stabilized
compliance-icons compliance-icons
Product Identifier
  • 1,1,1,3,3,3-Hexafluoroisopropyl methacrylate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : 1,1,1,3,3,3-Hexafluoroisopropyl methacrylate