missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
BDNF Monoclonal antibody specifically detects BDNF in Human samples. It is validated for Flow Cytometry, CyTOF
Specifications
Specifications
| Antigen | BDNF |
| Applications | Flow Cytometry, CyTOF |
| Classification | Monoclonal |
| Clone | 35909 |
| Conjugate | Janelia Fluor 646 |
| Dilution | Intracellular Staining by Flow Cytometry, CyTOF-ready |
| Formulation | 50mM Sodium Borate |
| Gene Alias | abrineurin, brain-derived neurotrophic factor, MGC34632, neurotrophin |
| Host Species | Mouse |
| Immunogen | S. frugiperda insect ovarian cell line Sf 21-derived recombinant human BDNF, His129-Arg247, Accession # P23560 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?