missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Naphthol-1,3,4,5,6,7,8-d7, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.50g
Specifications
Specifications
| Chemical Name or Material | 2-Naphthol (ring-d7) |
| CAS | 78832-54-9 |
| Molecular Formula | C10 2H7 H O |
| Quantity | 0.25 g |
| Synonym | 2-Naphthalen-1,3,4,5,6,7,8-d7-ol, 2-Naphthol-d7, 2-Naphthol (ring-D7) |
| InChI Formula | InChI=1 S/C10H8O/c11-10-6-5-8-3-1-2-4-9(8)7-10/h1-7,11 H/i1D,2 D,3 D,4 D,5 D,6 D,7 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c2c([2 H])c(O)c([2 H])c([2 H])c2c1[2 H] |
| IUPAC Name | 1,3,4,5,6,7,8-heptadeuterionaphthalen-2-ol |
| Molecular Weight (g/mol) | 151.21 |
| Formula Weight | 151.1015 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction