Learn More
Axygen™ 0.2 mL Polypropylene PCR Tube Strips
Product Code. 16282591 Shop All Axygen Scientific Products
Change view
Click to view available options
Closure Type:
Domed Cap
Flat Cap
Unit Size:
Pack of 1260
Product Code. Closure Type unitSize
16282591 Flat Cap Pack of 1260
16272591 Domed Cap Pack of 1260
2 options
Product Code. 16282591
Brand: Axygen™ PCR0208AFC
Available in a variety of colors for easy sample identification. Axygen™ 8-Strip PCR Tubes, 0.2 mL are constructed of sturdy polypropylene. Each non-sterile, conical tube features thin and consistent wall thickness. Sold with flat or domed caps to prevent sample loss or without caps.
Axygen™ 0.2 mL thin-wall 8 strip PCR tubes, available in an assortment of colors are designed to fit a wide variety of thermal cyclers. Ultra-thin and consistent wall thickness allows precise thermal transfer, even insertion cap ensures a tight fit to prevent evaporation.
  • RNase-/DNase-free
  • Nonpyrogenic
  • non-sterile
  • Manufactured with 99.9% pure virgin polypropylene
  • Caps are included
TRUSTED_SUSTAINABILITY
Specifications
Color Clear
Volume (Metric) 0.2 mL
Quantity 1250 Strips (8 Tubes/Strip)
Sterility Non-sterile
Closure Type Flat Cap