Learn More
HIF-2 alpha/EPAS1 Antibody (2597C), CoraFluor™ 1, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals (Bio-Techne) NBP3-07075CL1
Product Code. 30153256
  • 9225.00 NOK / 0.10mL
Description
HIF-2 alpha/EPAS1 Monoclonal antibody specifically detects HIF-2 alpha/EPAS1 in Human,Mouse samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
HIF-2 alpha/EPAS1
Monoclonal
CoraFluor 1
Basic-helix-loop-helix-PAS protein MOP2, BHLHE73, Class E basic helix-loop-helix protein 73, ECYT4, endothelial PAS domain protein 1, endothelial PAS domain-containing protein 1, EPAS1, EPAS-1, HIF-1-alpha-like factor, HIF-1alpha-like factor, HIF-2 alpha, HIF2A, hif2a angiogenesis, HIF-2-alpha, HIF2-alpha, HLF, hypoxia-inducible factor 2 alpha, Hypoxia-inducible factor 2-alpha, Member of PAS protein 2, MOP2, PAS domain-containing protein 2, PASD2
Partial recombinant mouse HIF-2 alpha/EPAS1 (amino acids 542-874) [UniProt Q6PEU2]
0.1 mL
Angiogenesis, Apoptosis, Cancer, Cancer Stem Cells, Chromatin Research, HIF Target Genes, Hypoxia, Lipid and Metabolism, Transcription Factors and Regulators
2034
Store at 4°C in the dark. Do not freeze.
IgG
Immunohistochemistry (Paraffin)
2597C
PBS
Rabbit
Protein A or G purified
RUO
Primary
Human, Mouse