missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Biotin Antibody (BTN/8672R), DyLight 488, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-20861G
This item is not returnable.
View return policy
Description
Biotin Monoclonal antibody specifically detects Biotin in All samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), In-situ Hybridization
Specifications
| Biotin | |
| Monoclonal | |
| DyLight 488 | |
| Vitamin B4 | |
| Biotinylated sheep immunoglobulin | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG κ |
| Western Blot, Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), In Situ Hybridization (ISH) | |
| BTN/8672R | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| All species | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction