Learn More
beta Amyloid Antibody (U.Illinois scFv59), CoraFluor™ 1, Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals NBP3-28660CL1
Product Code. 30141162
  • 7855.00 NOK / 0.10mL
Description
beta Amyloid Monoclonal antibody specifically detects beta Amyloid in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
beta Amyloid
Monoclonal
CoraFluor 1
Abeta, Amyloid beta, beta-amyloid peptide
Amyloid Beta
0.1 mL
Alzheimers Research, Apoptosis, Cardiovascular Biology, Cell Cycle and Replication, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Phospho Specific, Signal Transduction
351
Store at 4°C in the dark. Do not freeze.
IgG1
Flow Cytometry, ELISA, Functional Assay
U.Illinois scFv59
PBS
Human
Protein A purified