Learn More
0.1 NTU Ratio Turbidity Standard, Reagecon
BIOPHARMA
Product Code. 11429159 Shop All Reagecon Products
Click to view available options
Volume (Metric):
500 mL
Unit Size:
500mL
Product Code. 11429159
Brand: Reagecon CRSR0.1500
A suspension of polymer microspheres designed to overcome the adverse safety, accuracy and stability issues experienced with other turbidity standards
  • Guaranteed stability throughout shelf life even after opening bottle
  • ± 1% accuracy
  • NIST traceable
  • US EPA approved
Specifications
For Use With (Equipment) Ratio Instruments
Form Liquid
Accuracy ± 1% NTU
Volume (Metric) 500 mL
Packaging Type HDPE Bottle GL45