siRNA Details
| Gene Symbol: Dpp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012789.1 | NP_036921.1 | 13 | 1196 | ||
| XM_008761879.1 | XP_008760101.1 | 14 | 1705 | |||
| GenBank mRNA | J02997.1 | - | 13 | 1136 | ||
| J04591.1 | - | 13 | 1196 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dpp4| Gene Symbol: Dpp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012789.1 | NP_036921.1 | 13 | 1196 | ||
| XM_008761879.1 | XP_008760101.1 | 14 | 1705 | |||
| GenBank mRNA | J02997.1 | - | 13 | 1136 | ||
| J04591.1 | - | 13 | 1196 | |||