accessibility menu, dialog, popup

UserName

CAS RN 98027-84-0

NClClI

CAS RN 98027-84-0

IUPAC Name: 2,6-dichloro-4-iodopyridine
Synonyms: 2,6-dichloro-4-iodopyridine
Molecular Weight (g/mol): 273.88
Molecular Formula: C5H2Cl2IN
InChi Key: NGSKFMPSBUAUNE-UHFFFAOYSA-N
SMILES: ClC1=CC(I)=CC(Cl)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results