accessibility menu, dialog, popup

Test

CAS RN 97-08-5

CAS RN 97-08-5

IUPAC Name: 4-chloro-3-nitrobenzene-1-sulfonyl chloride
Synonyms: 4-chloro-3-nitrobenzenesulfonyl chloride
Molecular Weight (g/mol): 256.05
Molecular Formula: C6H3Cl2NO4S
InChi Key: SEWNAJIUKSTYOP-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(=CC=C1Cl)S(Cl)(=O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)