accessibility menu, dialog, popup

Test

CAS RN 96530-81-3

CAS RN 96530-81-3

IUPAC Name: 4-fluoro-2-methoxypyridine
Synonyms: 4-fluoro-2-methoxypyridine
Molecular Weight (g/mol): 127.12
Molecular Formula: C6H6FNO
InChi Key: RGWFJFBUHTUVMV-UHFFFAOYSA-N
SMILES: COC1=NC=CC(F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results