CAS RN 96515-79-6
CAS RN 96515-79-6
IUPAC Name:
5-chloro-2-fluorobenzaldehyde
Synonyms:
5-chloro-2-fluorobenzaldehyde
Molecular Weight (g/mol):
158.56
Molecular Formula:
C7H4ClFO
InChi Key:
WDTUCEMLUHTMCB-UHFFFAOYSA-N
SMILES:
FC1=CC=C(Cl)C=C1C=O
1
–
1
of
1
results
1
5-Chloro-2-fluorobenzaldehyde, 97%, Thermo Scientific Chemicals
CAS: 96515-79-6 Molecular Formula: C7H4ClFO Molecular Weight (g/mol): 158.56 MDL Number: MFCD03094518 InChI Key: WDTUCEMLUHTMCB-UHFFFAOYSA-N Synonym: 2-fluoro-5-chlorobenzaldehyde,3-chloro-6-fluoro benzaldehyde,3-chloro-6-fluorobenzaldehyde,5-chloro-2-fluoro-benzaldehyde,benzaldehyde, 5-chloro-2-fluoro,2-fluoro-5-chloro-benzaldehyde,zlchem 333,pubchem1411,ksc486m0t,2-fluoro-5-chloro benzaldehyde PubChem CID: 2773586 IUPAC Name: 5-chloro-2-fluorobenzaldehyde SMILES: FC1=CC=C(Cl)C=C1C=O