accessibility menu, dialog, popup

UserName

ClS

CAS RN 96-43-5

IUPAC Name: 2-chlorothiophene
Synonyms: 2-chlorothiophene
Molecular Weight (g/mol): 118.58
Molecular Formula: C4H3ClS
InChi Key: GSFNQBFZFXUTBN-UHFFFAOYSA-N
SMILES: ClC1=CC=CS1
Molecular Weight (g/mol) 
  • (3)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (3)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)