accessibility menu, dialog, popup

Test

CAS RN 94790-35-9

CAS RN 94790-35-9

IUPAC Name: [bis(dimethylamino)methylidene]-λ³-chloranylium
Synonyms: [bis(dimethylamino)methylidene]-λ³-chloranylium
Molecular Weight (g/mol): 135.61
Molecular Formula: C5H12ClN2
InChi Key: QZFWNEASXBNLCD-UHFFFAOYSA-N
SMILES: CN(C)C(=[Cl+])N(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results