CAS RN 946409-40-1
CAS RN 946409-40-1
IUPAC Name:
1-[(pyridin-3-yl)methyl]piperidine-4-carboxylic acid dihydrochloride
Synonyms:
1-(pyridin-3-ylmethyl)piperidine-4-carboxylic acid dihydrochloride
Molecular Weight (g/mol):
293.19
Molecular Formula:
C12H18Cl2N2O2
InChi Key:
FLINLLCJYJQIIY-UHFFFAOYSA-N
SMILES:
Cl.Cl.OC(=O)C1CCN(CC2=CN=CC=C2)CC1
1
–
1
of
1
results
1
1-(pyrid-3-ylmethyl)piperidine-4-carboxylic acid dihydrochloride, 97%, Thermo Scientific™
CAS: 946409-40-1 Molecular Formula: C12H18Cl2N2O2 Molecular Weight (g/mol): 293.188 MDL Number: MFCD06801260 InChI Key: FLINLLCJYJQIIY-UHFFFAOYSA-N Synonym: 1-pyridin-3-ylmethyl piperidine-4-carboxylic acid dihydrochloride,1-pyrid-3-ylmethyl piperidine-4-carboxylic acid dihydrochloride,1-pyridin-3-yl methyl piperidine-4-carboxylic acid dihydrochloride,1-pyridin-3-yl methyl piperidine-4-carboxylic acid-hydrogen chloride 1/2 PubChem CID: 42935493 IUPAC Name: 1-(pyridin-3-ylmethyl)piperidine-4-carboxylic acid;dihydrochloride SMILES: C1CN(CCC1C(=O)O)CC2=CN=CC=C2.Cl.Cl