accessibility menu, dialog, popup

UserName

SBrOBr

CAS RN 944450-89-9

IUPAC Name: 2-bromo-1-[4-(3-bromothiophen-2-yl)phenyl]ethan-1-one
Synonyms: 2-bromo-1-[4-(3-bromothiophen-2-yl)phenyl]ethanone
Molecular Weight (g/mol): 360.06
Molecular Formula: C12H8Br2OS
InChi Key: YTVAARIMMJZZIE-UHFFFAOYSA-N
SMILES: BrCC(=O)C1=CC=C(C=C1)C1=C(Br)C=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results