accessibility menu, dialog, popup

UserName

CAS RN 942206-05-5

IUPAC Name: 6-bromo-3-iodo-2,4-dimethylpyridine
Synonyms: 6-bromo-3-iodo-2,4-dimethylpyridine
Molecular Weight (g/mol): 311.95
Molecular Formula: C7H7BrIN
InChi Key: DPAHMUFCBJVARG-UHFFFAOYSA-N
SMILES: CC1=CC(Br)=NC(C)=C1I

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results