CAS RN 94164-88-2
CAS RN 94164-88-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
PKM2-IN-1, MedChemExpress
MedChemExpress PKM2-IN-1 is a pyruvate kinase M2 (PKM2) inhibitor with an IC50 of 2.95 μM.
