accessibility menu, dialog, popup

UserName

CAS RN 94088-46-7

H3COFN

CAS RN 94088-46-7

IUPAC Name: 2-fluoro-6-methoxybenzonitrile
Synonyms: 2-fluoro-6-methoxybenzonitrile
Molecular Weight (g/mol): 151.14
Molecular Formula: C8H6FNO
InChi Key: YPMSIWYNTPSPMV-UHFFFAOYSA-N
SMILES: COC1=CC=CC(F)=C1C#N
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results