accessibility menu, dialog, popup

UserName

CAS RN 93972-02-2

IUPAC Name: 4-decyl-1,1'-biphenyl
Synonyms: 4-decyl-1,1'-biphenyl
Molecular Weight (g/mol): 294.48
Molecular Formula: C22H30
InChi Key: YYBDFGOLQQVHDV-UHFFFAOYSA-N
SMILES: CCCCCCCCCCC1=CC=C(C=C1)C1=CC=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results